missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Nitroacetate 95.0+%, TCI America™

Catalog No. p-7133914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06575G 5 g
N065725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06575G Supplier TCI America Supplier No. N06575G
Only null left

Chemical Identifiers

CAS 626-35-7
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00007403
InChI Key FTKASJMIPSSXBP-UHFFFAOYSA-N
Synonym ethyl nitroacetate, nitroacetic acid ethyl ester, acetic acid, nitro-, ethyl ester, nitro ethyl acetate, ethylnitroacetate, acetic acid, 2-nitro-, ethyl ester, ethylnitroacetat, ethyinitroacetate, ethyl-2-nitroacetate, zlchem 269
PubChem CID 69379
IUPAC Name ethyl 2-nitroacetate
SMILES CCOC(=O)C[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 107°C
Quantity 5 g
Formula Weight 133.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Nitroacetate
RTECSNumber AJ1074000
TSCA Yes