missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nitroacetate, 97%

Catalog No. p-7021903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1443303 1 g
AAA1443306 5 g
AAA1443314 25 g
AAA1443322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1443303 Supplier Thermo Scientific Chemicals Supplier No. A1443303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-35-7 | C4H7NO4 | 133.103 g/mol

Ethyl nitroacetate has been used in the synthesis of γ-oxoacids via Michael addition reaction with α,β-unsaturated ketones, fuctionalization of C4-position on pyrimidine and C6-position on 2-deoxyguanosine to produce novel nucleosides, facile synthesis of α,α-diisobutylglycine, synthesis of DL-4,4-difluoroglutamic acid. It is also used as an intermediate in the preparation of unsubstituted amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl nitroacetate has been used in the synthesis of γ-oxoacids via Michael addition reaction with α,β-unsaturated ketones, fuctionalization of C4-position on pyrimidine and C6-position on 2′-deoxyguanosine to produce novel nucleosides, facile synthesis of α,α-diisobutylglycine, synthesis of DL-4,4-difluoroglutamic acid. It is also used as an intermediate in the preparation of unsubstituted amino acids.

Solubility
Slightly soluble in water. Soluble in chloroform and ethyl acetate.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 626-35-7
Molecular Formula C4H7NO4
Molecular Weight (g/mol) 133.103
MDL Number MFCD00007403
InChI Key FTKASJMIPSSXBP-UHFFFAOYSA-N
Synonym ethyl nitroacetate, nitroacetic acid ethyl ester, acetic acid, nitro-, ethyl ester, nitro ethyl acetate, ethylnitroacetate, acetic acid, 2-nitro-, ethyl ester, ethylnitroacetat, ethyinitroacetate, ethyl-2-nitroacetate, zlchem 269
PubChem CID 69379
IUPAC Name ethyl 2-nitroacetate
SMILES CCOC(=O)C[N+](=O)[O-]

Specifications

Density 1.202
Boiling Point 93°C to 95°C (10 mmHg)
Flash Point 92°C (197°F)
Odor Weak
Refractive Index 1.424
Linear Formula O2NCH2CO2CH2CH3
Quantity 1 g
Beilstein 1210027
Solubility Information Slightly soluble in water. Soluble in chloroform and ethyl acetate.
Formula Weight 133.11
Percent Purity 97%
Chemical Name or Material Ethyl nitroacetate
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-944-7
RTECSNumber AJ1074000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only