missing translation for 'onlineSavingsMsg'
Learn More

Ethyl nicotinate, 99%

Catalog No. AAL0321536
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0321536 500 g
AAL0321522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0321536 Supplier Thermo Scientific Chemicals Supplier No. L0321536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 614-18-6 | C8H9NO2 | 151.165 g/mol

Ethyl nicotinate is used for skin-conditioning cosmetics. It is principally used in medicine as a local vasodilator at a concentration of 1 to 2 % in creams, ointments and pomades. It is utilized for the treatment sprains, wrenches, muscular pains and tendonitis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl nicotinate is used for skin-conditioning cosmetics. It is principally used in medicine as a local vasodilator at a concentration of 1 to 2 % in creams, ointments and pomades. It is utilized for the treatment sprains, wrenches, muscular pains and tendonitis.

Solubility
Miscible with water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 614-18-6
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00006389
InChI Key XBLVHTDFJBKJLG-UHFFFAOYSA-N
Synonym ethyl nicotinate, ethyl nicotinoate, nicotinic acid ethyl ester, ethyl 3-pyridinecarboxylate, 3-pyridinecarboxylic acid, ethyl ester, ethylnicotinate, nicotinic acid, ethyl ester, mucotherm, nicaethan, nikethan
PubChem CID 69188
IUPAC Name ethyl pyridine-3-carboxylate
SMILES CCOC(=O)C1=CN=CC=C1

Specifications

Melting Point 8°C to 9°C
Density 1.107
Boiling Point 223°C to 224°C
Flash Point 93°C (199°F)
Odor Characteristic
Refractive Index 1.503
Quantity 500 g
Beilstein 122937
Solubility Information Miscible with water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material Ethyl nicotinate
Show More Show Less
Hazard Category H227-H315-H319-H500
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-370-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only