missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl N-(tert-Butoxycarbonyl)-D-pyroglutamate 95.0+%, TCI America™
Supplier: TCI America E09401G
Specifications
| Ethyl N-(tert-Butoxycarbonyl)-D-pyroglutamate | |
| 56°C | |
| C12H19NO5 | |
| 1 g | |
| YWWWGFSJHCFVOW-MRVPVSSYSA-N | |
| 1-O-tert-butyl 2-O-ethyl (2R)-5-oxopyrrolidine-1,2-dicarboxylate | |
| 820885 | |
| ≥95.0% (GC) |
| 144978-35-8 | |
| White | |
| MFCD09261329 | |
| N-(tert-Butoxycarbonyl)-D-pyroglutamic Acid Ethyl Ester, Ethyl (R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, (R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic Acid Ethyl Ester, Ethyl N-Boc-D-pyroglutamate, N-Boc-D-pyroglutamic Acid E | |
| CCOC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C | |
| 257.286 | |
| 257.29 | |
| Crystalline Powder |
Chemical Identifiers
| 144978-35-8 | |
| 257.286 | |
| YWWWGFSJHCFVOW-MRVPVSSYSA-N | |
| 820885 | |
| CCOC(=O)C1CCC(=O)N1C(=O)OC(C)(C)C |
| C12H19NO5 | |
| MFCD09261329 | |
| N-(tert-Butoxycarbonyl)-D-pyroglutamic Acid Ethyl Ester, Ethyl (R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, (R)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic Acid Ethyl Ester, Ethyl N-Boc-D-pyroglutamate, N-Boc-D-pyroglutamic Acid E | |
| 1-O-tert-butyl 2-O-ethyl (2R)-5-oxopyrrolidine-1,2-dicarboxylate |
Safety and Handling
TSCA : No