missing translation for 'onlineSavingsMsg'
Learn More

Ethyl N-Phenylformimidate 98.0+%, TCI America™

Catalog No. E04955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04955G 5 g
E049525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04955G Supplier TCI America Supplier No. E04955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6780-49-0
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00043655
InChI Key DRDBNKYFCOLNQO-UHFFFAOYSA-N
Synonym ethyl n-phenylformimidate, ethyl isoformanilide, n-ethoxymethyleneaniline, methanimidic acid, n-phenyl-, ethyl ester, ethyl phenylimidoformate, n-ethoxymethylene aniline, ethyl n-phenylcarboximidate, ethylisoformanilide, e-ethyl n-phenylcarboximidate, ethyl iso-formanilide
PubChem CID 111236
IUPAC Name ethyl N-phenylcarboximidate
SMILES CCOC=NC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 215°C
Quantity 5 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl N-Phenylformimidate