missing translation for 'onlineSavingsMsg'
Learn More

Ethyl n-Octanoate 98.0+%, TCI America™

Catalog No. p-7134140
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O003025ML 25 mL
O0030500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O003025ML Supplier TCI America Supplier No. O003025ML
Only null left

Chemical Identifiers

CAS 106-32-1
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00009552
InChI Key YYZUSRORWSJGET-UHFFFAOYSA-N
Synonym ethyl caprylate, octanoic acid, ethyl ester, ethyl n-octanoate, caprylic acid ethyl ester, ethyl octoate, ethyl octylate, octanoic acid ethyl ester, natural, unii-81c5mop582, fema no. 2449
PubChem CID 7799
ChEBI CHEBI:87426
IUPAC Name ethyl octanoate
SMILES CCCCCCCC(=O)OCC

Specifications

Melting Point -43°C
Color Yellow
Boiling Point 208°C
Quantity 25 mL
Formula Weight 172.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl n-Octanoate
EINECSNumber (2)-0782
RTECSNumber RH0680000
TSCA Yes