missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methyl Sulfone 98.0+%, TCI America™

Catalog No. p-7129019
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E047425G 25 g
E0474100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E047425G Supplier TCI America Supplier No. E047425G

Chemical Identifiers

CAS 594-43-4
Molecular Formula C3H8O2S
Molecular Weight (g/mol) 108.16
MDL Number MFCD00015543
InChI Key YBJCDTIWNDBNTM-UHFFFAOYSA-N
Synonym Methylsulfonylethane
PubChem CID 79059
IUPAC Name methanesulfonylethane
SMILES CCS(C)(=O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 108.16
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Ethyl Methyl Sulfone