missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methyl Carbonate 98.0+%, TCI America™

Catalog No. p-7124926
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C134210ML 10 mL
C134225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C134210ML Supplier TCI America Supplier No. C134210ML
Only null left

Chemical Identifiers

CAS 623-53-0
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00191398
InChI Key JBTWLSYIZRCDFO-UHFFFAOYSA-N
Synonym Carbonic Acid Ethyl Methyl Ester
PubChem CID 522046
IUPAC Name ethyl methyl carbonate
SMILES CCOC(=O)OC

Specifications

Color Colorless
Boiling Point 107°C
Quantity 10 mL
UN Number 3272
Formula Weight 104.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Methyl Carbonate