missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Methacrylate (stabilized with HQ) 99.0+%, TCI America™

Catalog No. M008425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M008425ML 25 mL
M0084500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M008425ML Supplier TCI America Supplier No. M008425ML

Chemical Identifiers

CAS 97-63-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00009161
InChI Key SUPCQIBBMFXVTL-UHFFFAOYSA-N
Synonym ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester
PubChem CID 7343
IUPAC Name ethyl 2-methylprop-2-enoate
SMILES CCOC(=O)C(=C)C

Specifications

Melting Point -75°C
Color Colorless
Boiling Point 117°C
Quantity 25 mL
UN Number 2277
Formula Weight 114.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Methacrylate (stabilized with HQ)
EINECSNumber (2)-1039
RTECSNumber OZ4550000
TSCA Yes