missing translation for 'onlineSavingsMsg'
Learn More

Ethyl methacrylate, 98+%, stab. with 10-120ppm, 4-methoxyphenol

Catalog No. p-7023700
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17518AK 250 mL
AAA17518AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA17518AK Supplier Thermo Scientific Chemicals Supplier No. A17518AK
Only null left

CAS: 97-63-2 | C6H10O2 | 114.144 g/mol

Ethyl methacrylate is used to prepare copolymers of EMA on waxy maize starch and hydroxypropylstarch, potential application of these copolymers are as excipients for compressed non-disintegrating matrix tablets. It is also used to study reactions of n-butyl acrylate and EMA with ozone in the gas phase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl methacrylate is used to prepare copolymers of EMA on waxy maize starch and hydroxypropylstarch, potential application of these copolymers are as excipients for compressed non-disintegrating matrix tablets. It is also used to study reactions of n-butyl acrylate and EMA with ozone in the gas phase.

Solubility
Soluble in water (4.69g/L), ether, and alcohols.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 97-63-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00009161
InChI Key SUPCQIBBMFXVTL-UHFFFAOYSA-N
Synonym ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester
PubChem CID 7343
IUPAC Name ethyl 2-methylprop-2-enoate
SMILES CCOC(=O)C(=C)C

Specifications

Melting Point -60°C
Density 0.917
Boiling Point 118°C to 119°C
Flash Point 18°C (64°F)
Odor Pungent
Refractive Index 1.414
Quantity 250 mL
UN Number UN2277
Beilstein 471201
Solubility Information Soluble in water (4.69g/L),ether,and alcohols.
Formula Weight 114.15
Percent Purity ≥98%
Chemical Name or Material Ethyl methacrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less
Hazard Category H225-H315-H317-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H317-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL METHACRYLATE, STABILIZED
EINECSNumber 202-597-5
RTECSNumber OZ4550000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only