missing translation for 'onlineSavingsMsg'
Learn More

Ethyl mesitylacetate, 97%

Catalog No. p-7044799
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0827504 2 g
AAL0827509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0827504 Supplier Thermo Scientific Chemicals Supplier No. L0827504
Only null left

CAS: 5460-08-2 | C13H18O2 | 206.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5460-08-2
MDL Number MFCD00051916

Specifications

Density 1.034
Boiling Point 135°C to 140°C (9 mmHg)
Molecular Formula C13H18O2
Refractive Index 1.505
Quantity 2 g
Beilstein 2105791
Synonym ethyl mesitylacetate, ethyl 2-2,4,6-trimethylphenyl acetate, benzeneacetic acid, 2,4,6-trimethyl-, ethyl ester, ethyl 2-mesitylacetate, acmc-20akx5, mesitylacetic acid ethyl ester, ethyl 2,4,6-trimethylphenylacetate, ethyl 2,4,6-trimethylphenyl acetate, 2,4,6-trimethyl-phenyl-acetic acid ethyl ester, benzeneacetic acid,2,4,6-trimethyl-, ethyl ester
InChI Key OFZZPFNYAMIBNU-UHFFFAOYSA-N
SMILES CCOC(=O)CC1=C(C=C(C=C1C)C)C
IUPAC Name ethyl 2-(2,4,6-trimethylphenyl)acetate
Molecular Weight (g/mol) 206.29
PubChem CID 222429
Formula Weight 206.29
Percent Purity 97%
Chemical Name or Material Ethyl mesitylacetate
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only