missing translation for 'onlineSavingsMsg'
Learn More

Ethyl mercaptoacetate, 98+%

Catalog No. p-7021826
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1432106 5 g
AAA1432122 100 g
AAA1432136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1432106 Supplier Thermo Scientific Chemicals Supplier No. A1432106
Only null left

CAS: 623-51-8 | C4H8O2S | 120.166 g/mol

It finds its application in the reaction of the dilithio-derivative with an aldehyde, followed by conversion to the episulfide by reaction with ethyl chloroformate, and desulfurization with triethyl phosphite, leads to the (E)-2-alkenoic acid with high stereoselectivity. It is used as depilatories, hair straightening agent, hair waving agent, and reducing agent in cosmetic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the reaction of the dilithio-derivative with an aldehyde, followed by conversion to the episulfide by reaction with ethyl chloroformate, and desulfurization with triethyl phosphite, leads to the (E)-2-alkenoic acid with high stereoselectivity. It is used as depilatories, hair straightening agent, hair waving agent, and reducing agent in cosmetic industry.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and store away from ignition sources. Store under inert gas.

Chemical Identifiers

CAS 623-51-8
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.166
MDL Number MFCD00004874
InChI Key PVBRSNZAOAJRKO-UHFFFAOYSA-N
Synonym ethyl thioglycolate, ethyl mercaptoacetate, ethyl 2-mercaptoacetate, mercaptoacetic acid ethyl ester, ethyl thioglycollate, acetic acid, mercapto-, ethyl ester, thioglycolic acid ethyl ester, ethyl mercaptoacetic acid, usaf ek-2070, acetic acid, 2-mercapto-, ethyl ester
PubChem CID 12185
IUPAC Name ethyl 2-sulfanylacetate
SMILES CCOC(=O)CS

Specifications

Melting Point -80°C
Density 1.096
Boiling Point 156°C to 159°C
Flash Point 48°C (118°F)
Odor Stench
Refractive Index 1.458
Linear Formula HSCH2CO2CH2CH3
Quantity 5 g
UN Number UN1228
Beilstein 605581
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 120.17
Percent Purity ≥98%
Chemical Name or Material Ethyl mercaptoacetate
Show More Show Less
Hazard Category H226-H301-H315-H319
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P305+P351+P338-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, TOXIC, N.O.S.
EINECSNumber 210-800-3
RTECSNumber AI6650000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only