missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Mercaptan 98.0+%, TCI America™

Catalog No. p-7128798
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E003625ML 25 mL
E0036500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E003625ML Supplier TCI America Supplier No. E003625ML
Only null left

Chemical Identifiers

CAS 75-08-1
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00004887
InChI Key DNJIEGIFACGWOD-UHFFFAOYSA-N
Synonym ethyl mercaptan, mercaptoethane, thioethanol, ethyl hydrosulfide, ethyl sulfhydrate, ethyl thioalcohol, thioethyl alcohol, 1-mercaptoethane, aethanethiol, aethylmercaptan
PubChem CID 6343
IUPAC Name ethanethiol
SMILES CCS

Specifications

Color Colorless
Boiling Point 35°C
Quantity 25 mL
UN Number 2363
Formula Weight 62.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Mercaptan
EINECSNumber (2)-0460
RTECSNumber KI9625000
TSCA Yes