missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Maltol, FCC, 99%, Spectrum™ Chemical
E1061, 4940-11-8, C7H8O3
$990.77 - $990.77
Chemical Identifiers
| CAS | 11-8-4940 |
|---|---|
| Molecular Formula | C7H8O3 |
| Molecular Weight (g/mol) | 140.14 |
| InChI Key | YIKYNHJUKRTCJL-UHFFFAOYSA-N |
| IUPAC Name | 2-ethyl-3-hydroxy-4H-pyran-4-one |
| SMILES | CCC1=C(O)C(=O)C=CO1 |