missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Malonyl Chloride 97.0+%, TCI America™

Catalog No. p-7129024
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04845G 5 g
E048425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04845G Supplier TCI America Supplier No. E04845G
Only null left

Chemical Identifiers

CAS 36239-09-5
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00000736
InChI Key KWFADUNOPOSMIJ-UHFFFAOYSA-N
Synonym ethyl 3-chloro-3-oxopropionate, ethyl chloroformylacetate, ethyl chloroformyl acetate, propanoic acid, 3-chloro-3-oxo-, ethyl ester, ethyl3-chloro-3-oxopropanoate, ethyl malonylchloride, carbethoxyacetyl chloride, 2-ethoxycarbonylacetyl chloride, ethyl 2-chlorocarbonyl acetate, chloroformyl acetic acid ethyl ester
PubChem CID 118931
IUPAC Name ethyl 3-chloro-3-oxopropanoate
SMILES CCOC(=O)CC(Cl)=O

Specifications

Color Yellow
Boiling Point 83°C
Quantity 5 g
UN Number 3265
Formula Weight 150.56
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl Malonyl Chloride