missing translation for 'onlineSavingsMsg'
Learn More

Ethyl malonyl chloride, 95%

Catalog No. AAA1561622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1561622 100 g
AAA1561606 5 g
AAA1561614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1561622 Supplier Thermo Scientific Chemicals Supplier No. A1561622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36239-09-5 | C5H7ClO3 | 150.56 g/mol

Ethyl malonyl chloride is used in the syntheses of methanofullerodendimers and 3-pyrrolin-2-ones. It plays a vital role in the preparation of 3,5-disubstituted 1,2,4-oxadiazole derivatives, which are potential peptidomimetic building blocks. It is a versatile acylating agent for propargyl alcohols, hydrazines and amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl malonyl chloride is used in the syntheses of methanofullerodendimers and 3-pyrrolin-2-ones. It plays a vital role in the preparation of 3,5-disubstituted 1,2,4-oxadiazole derivatives, which are potential peptidomimetic building blocks. It is a versatile acylating agent for propargyl alcohols, hydrazines and amines.

Solubility
Miscible with water.

Notes
Store in cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols. It reacts with water.

Chemical Identifiers

CAS 36239-09-5
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00000736
InChI Key KWFADUNOPOSMIJ-UHFFFAOYSA-N
Synonym ethyl 3-chloro-3-oxopropionate, ethyl chloroformylacetate, ethyl chloroformyl acetate, propanoic acid, 3-chloro-3-oxo-, ethyl ester, ethyl3-chloro-3-oxopropanoate, ethyl malonylchloride, carbethoxyacetyl chloride, 2-ethoxycarbonylacetyl chloride, ethyl 2-chlorocarbonyl acetate, chloroformyl acetic acid ethyl ester
PubChem CID 118931
IUPAC Name ethyl 3-chloro-3-oxopropanoate
SMILES CCOC(=O)CC(Cl)=O

Specifications

Density 1.189
Boiling Point 83°C (22 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.429
Quantity 100 g
UN Number UN3265
Beilstein 636215
Sensitivity Moisture sensitive
Solubility Information Miscible with water.
Formula Weight 150.56
Percent Purity 95%
Chemical Name or Material Ethyl malonyl chloride
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 252-934-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only