missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl levulinate, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Acidity | 5mg KOH/g max. |
| CAS | 539-88-8 |
| Density | 1.0120g/mL |
| Boiling Point | 206°C |
| Flash Point | 90°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Molecular Formula | C7H12O3 |
| Refractive Index | 1.4210 to 1.423 |
| Show More |
Safety and Handling
| EINECSNumber | 208-728-2 |
| RTECSNumber | OI1700000 |
| TSCA | TSCA |
RUO – Research Use Only