missing translation for 'onlineSavingsMsg'
Learn More

Ethyl levulinate, 98%

Catalog No. AAA1500118
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1500118 50 g
AAA1500122 100 g
AAA1500136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1500118 Supplier Thermo Scientific Chemicals Supplier No. A1500118

CAS: 539-88-8 | C7H12O3 | 144.17 g/mol

Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl levulinate is used to develop a preparative HPLC method to preserve wine aromas and isolates fruity characteristics of red wine aroma extracts

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 539-88-8
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009209
InChI Key GMEONFUTDYJSNV-UHFFFAOYSA-N
Synonym ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate
PubChem CID 10883
IUPAC Name ethyl 4-oxopentanoate
SMILES CCOC(=O)CCC(C)=O

Specifications

Density 1.014
Boiling Point 203°C to 205°C
Flash Point 94°C (201°F)
Refractive Index 1.422
Quantity 50 g
Beilstein 507641
Merck Index 14,3819
Solubility Information Soluble in water.
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material Ethyl levulinate
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 208-728-2
RTECSNumber OI1700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only