missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L-nipecotate, 97%

Catalog No. p-7038266
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAH5718503 1 g
AAH5718506 5 g
2 options

Catalog No. AAH5718503

Supplier: Thermo Scientific Chemicals H5718503

Only null left

CAS: 37675-18-6 | C8H15NO2 | 157.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37675-18-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00792499
InChI Key XIWBSOUNZWSFKU-ZETCQYMHSA-N
Synonym s-ethyl piperidine-3-carboxylate, ethyl 3s-piperidine-3-carboxylate, ethyl s-nipecotate, s-+-nipecotic acid ethyl ester, s-ethyl nipecotate, ethyl s-piperidine-3-carboxylate, 3-piperidinecarboxylic acid, ethyl ester, 3s, s-+-ethyl nipecotate, ethyl s-+-nipecotate, s-nipecotic acid ethyl ester
PubChem CID 187784
IUPAC Name ethyl (3S)-piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1

Specifications

Density 1.043
Flash Point 76°C (169°F)
Refractive Index 1.47
Quantity 1 g
Sensitivity Air, light, and moisture sensitive
Optical Rotation +1.5° (Neat)
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material Ethyl L-nipecotate
Hazard Category H227-H315-H318-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only