missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L-nipecotate, 97%

Catalog No. AAH5718506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5718506 5 g
AAH5718503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5718506 Supplier Thermo Scientific Chemicals Supplier No. H5718506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 37675-18-6 | C8H15NO2 | 157.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37675-18-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00792499
InChI Key XIWBSOUNZWSFKU-ZETCQYMHSA-N
Synonym s-ethyl piperidine-3-carboxylate, ethyl 3s-piperidine-3-carboxylate, ethyl s-nipecotate, s-+-nipecotic acid ethyl ester, s-ethyl nipecotate, ethyl s-piperidine-3-carboxylate, 3-piperidinecarboxylic acid, ethyl ester, 3s, s-+-ethyl nipecotate, ethyl s-+-nipecotate, s-nipecotic acid ethyl ester
PubChem CID 187784
IUPAC Name ethyl (3S)-piperidine-3-carboxylate
SMILES CCOC(=O)C1CCCNC1

Specifications

Density 1.043
Flash Point 76°C (169°F)
Refractive Index 1.47
Quantity 5 g
Sensitivity Air, light, and moisture sensitive
Optical Rotation +1.5° (Neat)
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material Ethyl L-nipecotate
Hazard Category H227-H315-H318-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only