missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl L-(-)-Lactate 98.0+%, TCI America™
$34.66 - $94.33
Chemical Identifiers
| CAS | 687-47-8 |
|---|---|
| Molecular Formula | C5H10O3 |
| Molecular Weight (g/mol) | 118.132 |
| MDL Number | MFCD00004518 |
| InChI Key | LZCLXQDLBQLTDK-BYPYZUCNSA-N |
| Synonym | ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s |
| PubChem CID | 92831 |
| ChEBI | CHEBI:78322 |
| IUPAC Name | ethyl (2S)-2-hydroxypropanoate |
| SMILES | CCOC(=O)C(C)O |
Chemical Identifiers
| 687-47-8 | |
| 118.132 | |
| LZCLXQDLBQLTDK-BYPYZUCNSA-N | |
| 92831 | |
| ethyl (2S)-2-hydroxypropanoate |
| C5H10O3 | |
| MFCD00004518 | |
| ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s | |
| CHEBI:78322 | |
| CCOC(=O)C(C)O |
Specifications
| 687-47-8 | |
| 70°C | |
| MFCD00004518 | |
| 1192 | |
| LZCLXQDLBQLTDK-BYPYZUCNSA-N | |
| ethyl (2S)-2-hydroxypropanoate | |
| 92831 | |
| 118.13 | |
| Liquid |
| Colorless | |
| C5H10O3 | |
| 25 g | |
| ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s | |
| CCOC(=O)C(C)O | |
| 118.132 | |
| CHEBI:78322 | |
| ≥98.0% (GC) | |
| Ethyl L-(-)-Lactate |
Safety and Handling
EINECSNumber : (2)-1371
RTECSNumber : OD5075000
TSCA : Yes