missing translation for 'onlineSavingsMsg'
Learn More

Ethyl L-(-)-Lactate 98.0+%, TCI America™

Catalog No. L016225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L016225G 25 g
L0162500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L016225G Supplier TCI America Supplier No. L016225G

Chemical Identifiers

CAS 687-47-8
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00004518
InChI Key LZCLXQDLBQLTDK-BYPYZUCNSA-N
Synonym ethyl l-lactate, ethyl 2s-2-hydroxypropanoate, l-ethyl lactate, l---ethyl lactate, ethyl l--lactate, ethyl s-2-hydroxypropionate, ethyl l---lactate, s-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s
PubChem CID 92831
ChEBI CHEBI:78322
IUPAC Name ethyl (2S)-2-hydroxypropanoate
SMILES CCOC(=O)C(C)O

Specifications

Color Colorless
Boiling Point 70°C
Quantity 25 g
UN Number 1192
Formula Weight 118.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl L-(-)-Lactate
EINECSNumber (2)-1371
RTECSNumber OD5075000
TSCA Yes