missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isovalerate 99.0+%, TCI America™

Catalog No. p-7131265
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I019425ML 25 mL
I0194500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I019425ML Supplier TCI America Supplier No. I019425ML
Only null left

Chemical Identifiers

CAS 108-64-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009203
InChI Key PPXUHEORWJQRHJ-UHFFFAOYSA-N
Synonym ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
PubChem CID 7945
ChEBI CHEBI:31571
IUPAC Name ethyl 3-methylbutanoate
SMILES CCOC(=O)CC(C)C

Specifications

Color Colorless
Boiling Point 134°C
Quantity 25 mL
UN Number 3272
Formula Weight 130.19
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Isovalerate
EINECSNumber (2)-0776
RTECSNumber NY1504000
TSCA Yes