missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isovalerate, 98%

Catalog No. AAA10292AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA10292AP 500 mL
AAA10292AE 100 mL
AAA102920F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA10292AP Supplier Thermo Scientific Chemicals Supplier No. A10292AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 108-64-5 | C7H14O2 | 130.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-64-5
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009203
InChI Key PPXUHEORWJQRHJ-UHFFFAOYSA-N
Synonym ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
PubChem CID 7945
ChEBI CHEBI:31571
IUPAC Name ethyl 3-methylbutanoate
SMILES CCOC(=O)CC(C)C

Specifications

Melting Point -99°C
Density 0.866
Boiling Point 131°C to 133°C
Flash Point 26°C (78°F)
Odor Fruit-like
Refractive Index 1.396
Quantity 500 mL
UN Number UN3272
Beilstein 1744677
Merck Index 14,3816
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material Ethyl isovalerate
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 203-602-3
RTECSNumber NY1504000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only