missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isonicotinoylacetate, 95%

Catalog No. AC425480025
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC425480025 2.5 g Glass bottle
AC425480100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC425480025 Supplier Thermo Scientific Chemicals Supplier No. 425480025
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 26377-17-3 | C10H11NO3 | 193.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26377-17-3
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.20
MDL Number MFCD00094021
InChI Key PCJNYGPKMQQCPX-UHFFFAOYSA-N
Synonym ethyl isonicotinoylacetate, ethyl 3-oxo-3-pyridin-4-yl propanoate, ethyl 3-oxo-3-4-pyridyl propionate, ethyl 3-4-pyridyl-3-oxopropionate, 3-oxo-3-pyridin-4-yl-propionic acid ethyl ester, ethyl 3-oxo-3-4-pyridinyl propanoate, ethylisonicotinoylacetate, ethyl-4-pyridoyl acetate, ethyl 3-oxo-3-4-pyridyl propanoate, 4-pyridinepropanoic acid, b-oxo-, ethyl ester
PubChem CID 2735202
IUPAC Name ethyl 3-oxo-3-pyridin-4-ylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=NC=C1

Specifications

Melting Point 53°C to 59°C
Color Brown-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 2.5 g
Formula Weight 193.2
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl isonicotinoylacetate
EINECSNumber 247-635-1

RUO – Research Use Only