missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isonicotinate 98.0+%, TCI America™

Catalog No. i0137500ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
I0137500ML 500 mL
I013725ML 25 mL
2 options

Catalog No. I0137500ML

Supplier: TCI America I0137500ML

Only null left

Chemical Identifiers

CAS 1570-45-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00006428
InChI Key MCRPKBUFXAKDKI-UHFFFAOYSA-N
Synonym ethyl isonicotinate, 4-picolinic acid ethyl ester, ethyl 4-pyridinecarboxylate, isonicotinic acid, ethyl ester, 4-carbethoxypyridine, 4-pyridinecarboxylic acid, ethyl ester, isonicotinic acid ethyl ester, 4-carboethoxypyridine, gamma-pyridinecarboxylic acid ethyl ester, .gamma.-pyridinecarboxylic acid ethyl ester
PubChem CID 15291
IUPAC Name ethyl pyridine-4-carboxylate
SMILES CCOC(=O)C1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 92°C
Quantity 500 mL
Formula Weight 151.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl Isonicotinate
EINECSNumber (5)-5677
RTECSNumber NS1450000
TSCA Yes