missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isonicotinate 98.0+%, TCI America™

Catalog No. I013725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I013725ML 25 mL
I0137500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I013725ML Supplier TCI America Supplier No. I013725ML

Chemical Identifiers

CAS 1570-45-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00006428
InChI Key MCRPKBUFXAKDKI-UHFFFAOYSA-N
Synonym ethyl isonicotinate, 4-picolinic acid ethyl ester, ethyl 4-pyridinecarboxylate, isonicotinic acid, ethyl ester, 4-carbethoxypyridine, 4-pyridinecarboxylic acid, ethyl ester, isonicotinic acid ethyl ester, 4-carboethoxypyridine, gamma-pyridinecarboxylic acid ethyl ester, .gamma.-pyridinecarboxylic acid ethyl ester
PubChem CID 15291
IUPAC Name ethyl pyridine-4-carboxylate
SMILES CCOC(=O)C1=CC=NC=C1

Specifications

Color Yellow
Boiling Point 92°C
Quantity 25 mL
Formula Weight 151.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl Isonicotinate
EINECSNumber (5)-5677
RTECSNumber NS1450000
TSCA Yes