missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isonicotinate, 98%

Catalog No. AAL0301022
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0301022 100 g
AAL0301036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0301022 Supplier Thermo Scientific Chemicals Supplier No. L0301022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1570-45-2 | C8H9NO2 | 151.165 g/mol

Ethyl isonicotinate is an organic building block, used in the synthesis of various pharmaceutical and biologically active compounds. It is also a pyridine compound that is shown to be able to trap the thrips in the corp.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl isonicotinate is an organic building block, used in the synthesis of various pharmaceutical and biologically active compounds. It is also a pyridine compound that is shown to be able to trap the thrips in the corp.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1570-45-2
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00006428
InChI Key MCRPKBUFXAKDKI-UHFFFAOYSA-N
Synonym ethyl isonicotinate, 4-picolinic acid ethyl ester, ethyl 4-pyridinecarboxylate, isonicotinic acid, ethyl ester, 4-carbethoxypyridine, 4-pyridinecarboxylic acid, ethyl ester, isonicotinic acid ethyl ester, 4-carboethoxypyridine, gamma-pyridinecarboxylic acid ethyl ester, .gamma.-pyridinecarboxylic acid ethyl ester
PubChem CID 15291
IUPAC Name ethyl pyridine-4-carboxylate
SMILES CCOC(=O)C1=CC=NC=C1

Specifications

Density 1.008
Boiling Point 220°C
Flash Point 87°C (188°F)
Odor Characteristic
Refractive Index 1.501
Quantity 100 g
Beilstein 122942
Merck Index 14,5187
Solubility Information Slightly soluble in water.
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material Ethyl isonicotinate
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 216-379-2
RTECSNumber NS1450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only