missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Isocyanatoacetate 97.0+%, TCI America™
$125.13 - $346.52
Chemical Identifiers
| CAS | 2949-22-6 |
|---|---|
| Molecular Formula | C5H7NO3 |
| Molecular Weight (g/mol) | 129.12 |
| MDL Number | MFCD00002040 |
| InChI Key | DUVOZUPPHBRJJO-UHFFFAOYSA-N |
| Synonym | ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester |
| PubChem CID | 76283 |
| IUPAC Name | ethyl 2-isocyanatoacetate |
| SMILES | CCOC(=O)CN=C=O |
Chemical Identifiers
| 2949-22-6 | |
| 129.12 | |
| DUVOZUPPHBRJJO-UHFFFAOYSA-N | |
| 76283 | |
| CCOC(=O)CN=C=O |
| C5H7NO3 | |
| MFCD00002040 | |
| ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester | |
| ethyl 2-isocyanatoacetate |
Specifications
| 2949-22-6 | |
| 80°C | |
| MFCD00002040 | |
| 2206 | |
| DUVOZUPPHBRJJO-UHFFFAOYSA-N | |
| ethyl 2-isocyanatoacetate | |
| 76283 | |
| ≥97.0% (GC) | |
| Ethyl Isocyanatoacetate |
| Yellow | |
| C5H7NO3 | |
| 5 g | |
| ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester | |
| CCOC(=O)CN=C=O | |
| 129.12 | |
| 129.12 | |
| Liquid |
Safety and Handling
RTECSNumber : AI4450000
TSCA : Yes