missing translation for 'onlineSavingsMsg'
Learn More

Ethyl isocyanatoacetate, 98%

CAS: 2949-22-6 | C5H7NO3 | 129.12 g/mol

Supplier:  Thermo Scientific Chemicals L1060906

 View more versions of this product

Catalog No. AAL1060906


Only null left
Explore available promotions

Description

Description

6-(carboxymethylureido)-(±)-nicotine (CMUNic)was prepared by using ethyl isocyanatoacetate. Ethyl isocyanatoacetate has been used in the preparation of nicotine immunogen, ethyl 8-carbamoyl-4-oxo-3,4-dihydroimidazo[5,1-d]-1,2,3,5-tetrazin-3-ylacetate, imidazo[1,2-c]-quinazoline-2,5-(3H,6H)dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-(carboxymethylureido)-(±)-nicotine (CMUNic)was prepared by using ethyl isocyanatoacetate. Ethyl isocyanatoacetate has been used in the preparation of nicotine immunogen, ethyl 8-carbamoyl-4-oxo-3,4-dihydroimidazo[5,1-d]-1,2,3,5-tetrazin-3-ylacetate, imidazo[1,2-c]-quinazoline-2,5-(3H,6H)dione.

Solubility
Reacts with water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, water, heat.
Specifications

Specifications

Ethyl isocyanatoacetate
1.151
72°C (161°F)
C5H7NO3
OCNCH2CO2CH2CH3
5 g
Moisture sensitive
Reacts with water.
CCOC(=O)CN=C=O
129.12
129.12
2949-22-6
66°C to 68°C (11 mmHg)
Pungent
1.424
MFCD00002040
970503
ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester
DUVOZUPPHBRJJO-UHFFFAOYSA-N
ethyl 2-isocyanatoacetate
76283
98%

Chemical Identifiers

2949-22-6
129.12
DUVOZUPPHBRJJO-UHFFFAOYSA-N
76283
CCOC(=O)CN=C=O
C5H7NO3
MFCD00002040
ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester
ethyl 2-isocyanatoacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H302-H335-H227-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c

H227-H302-H315-H317-H319-H334-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 220-967-4

RTECSNumber : AI4450000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only