missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Indole-3-carboxylate 97.0+%, TCI America™

Catalog No. p-7129245
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09551G 1 g
E09555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09551G Supplier TCI America Supplier No. E09551G

Chemical Identifiers

CAS 776-41-0
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
MDL Number MFCD00228454
InChI Key XOUHVMVYFOXTMN-UHFFFAOYSA-N
Synonym ethyl indole-3-carboxylate, indole-3-carboxylic acid ethyl ester, 1h-indole-3-carboxylic acid ethyl ester, 1h-indole-3-carboxylic acid, ethyl ester, indole3carboxylicacidethylester, pubchem7352, lindole-3-ethyl formate, 3-ethoxycarbonyl indole, ksc644o3d, 3-indolecarboxylicacidethylester
PubChem CID 247965
IUPAC Name ethyl 1H-indole-3-carboxylate
SMILES CCOC(=O)C1=CNC2=CC=CC=C21

Specifications

Melting Point 123°C
Quantity 1 g
Formula Weight 189.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Indole-3-carboxylate