missing translation for 'onlineSavingsMsg'
Learn More

Ethyl indole-3-acrylate, 97%

Catalog No. AAL0870303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0870303 1 g
AAL0870306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0870303 Supplier Thermo Scientific Chemicals Supplier No. L0870303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 15181-86-9 | C13H13NO2 | 215.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15181-86-9
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD00051947
InChI Key OQJSITNIWIYWPU-FPLPWBNLSA-N
Synonym ethyl 2z-3-1h-indol-3-yl prop-2-enoate
PubChem CID 13863197
IUPAC Name ethyl (Z)-3-(1H-indol-3-yl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CNC2=CC=CC=C21

Specifications

Melting Point 118°C to 122°C
Quantity 1 g
Beilstein 476317
Formula Weight 215.25
Percent Purity 97%
Chemical Name or Material Ethyl indole-3-acrylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only