missing translation for 'onlineSavingsMsg'
Learn More

Ethyl indole-3-acetate, 98+%

Catalog No. AAA1347406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1347406 5 g
AAA1347414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1347406 Supplier Thermo Scientific Chemicals Supplier No. A1347406

CAS: 778-82-5 | C12H13NO2 | 203.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 778-82-5
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00005635
InChI Key HUDBDWIQSIGUDI-UHFFFAOYSA-N
Synonym ethyl 3-indoleacetate, ethyl 2-1h-indol-3-yl acetate, indole-3-acetic acid ethyl ester, ethyl indole-3-acetate, ethyl indol-3-ylacetate, 1h-indole-3-acetic acid, ethyl ester, ethyl 1h-indol-3-ylacetate, indole-3-acetic acid, ethyl ester, ethyl beta-indolylacetate, ethyl3-indoleacetate
PubChem CID 13067
IUPAC Name ethyl 2-(1H-indol-3-yl)acetate
SMILES CCOC(=O)CC1=CNC2=CC=CC=C21

Specifications

Melting Point 41°C to 44°C
Boiling Point 164°C to 166°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 171682
Formula Weight 203.24
Percent Purity ≥98%
Chemical Name or Material Ethyl indole-3-acetate
Hazard Category H302+H312-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-296-0
RTECSNumber NL3528000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only