missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Indole-2-carboxylate 98.0+%, TCI America™

Catalog No. p-7131501
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I05855G 5 g
I058525G 25 g
2 options

Catalog No. I05855G

Supplier: TCI America I05855G

Only null left

Chemical Identifiers

CAS 3770-50-1
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005609
InChI Key QQXQAEWRSVZPJM-UHFFFAOYSA-N
Synonym ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole
PubChem CID 73125
IUPAC Name ethyl 1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC=C2N1

Specifications

Melting Point 122°C
Color White-Yellow
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Indole-2-carboxylate