missing translation for 'onlineSavingsMsg'
Learn More

Ethyl indole-2-carboxylate, 98+%

Catalog No. AAL0079706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0079706 5 g
AAL0079714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0079706 Supplier Thermo Scientific Chemicals Supplier No. L0079706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3770-50-1 | C11H11NO2 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3770-50-1
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005609
InChI Key QQXQAEWRSVZPJM-UHFFFAOYSA-N
Synonym ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole
PubChem CID 73125
IUPAC Name ethyl 1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC=C2N1

Specifications

Melting Point 122°C to 125°C
Quantity 5 g
Beilstein 146395
Formula Weight 189.21
Percent Purity ≥98%
Chemical Name or Material Ethyl indole-2-carboxylate
EINECSNumber 223-206-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only