missing translation for 'onlineSavingsMsg'
Learn More

Ethyl imidazole-2-carboxylate, 97%

Catalog No. AAL1940506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1940506 5 g
AAL1940503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1940506 Supplier Thermo Scientific Chemicals Supplier No. L1940506
Only null left

CAS: 33543-78-1 | C6H8N2O2 | 140.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33543-78-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD03426031
InChI Key UHYNYIGCGVDBTC-UHFFFAOYSA-N
Synonym ethyl imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid, ethyl ester, ethyl1h-imidazole-2-carboxylate, 1h-imidazole-2-carboxylic acid ethyl ester, ethyl 2-imidazolecarboxylate, imidazole-2-carboxylic acid ethyl ester, 2-ethoxycarbonyl-1h-imidazole, ethylimidazole-2-carboxylate, ethoxyformyl imidazole, zlchem 847
PubChem CID 549404
IUPAC Name ethyl 1H-imidazole-2-carboxylate
SMILES CCOC(=O)C1=NC=CN1

Specifications

Melting Point 176°C to 178°C
Boiling Point 48°C to 51°C (0.3 mmHg)
Quantity 5 g
Beilstein 115685
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material Ethyl imidazole-2-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only