missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrogen pimelate, 98%

Catalog No. AAL0402506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0402506 5 g
AAL0402514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0402506 Supplier Thermo Scientific Chemicals Supplier No. L0402506

CAS: 33018-91-6 | C9H16O4 | 188.223 g/mol

Ethyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 33018-91-6
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00801202
InChI Key NQYXFXWKKYGBNL-UHFFFAOYSA-N
Synonym ethyl hydrogen pimelate, pimelic acid monoethyl ester, heptanedioic acid monoethyl ester, ethyl hydrogen heptane-1,7-dioate, ethyl hydroden pimelate, heptandioic acid monoethyl ester, heptanedioic acid 1-ethyl ester, heptanedioic acid, 1-ethyl ester
PubChem CID 118383
IUPAC Name 7-ethoxy-7-oxoheptanoic acid
SMILES CCOC(=O)CCCCCC(=O)O

Specifications

Melting Point 10°C
Density 1.06
Boiling Point 135°C to 138°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.442
Linear Formula CH3CH2O2C(CH2)5CO2H
Quantity 5 g
Beilstein 1774583
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 188.22
Percent Purity 98%
Chemical Name or Material Ethyl hydrogen pimelate
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 251-346-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only