missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrogen malonate, 96%

Catalog No. AAA1262730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1262730 250 g
AAA1262709 10 g
AAA1262718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1262730 Supplier Thermo Scientific Chemicals Supplier No. A1262730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1071-46-1 | C5H8O4 | 132.115 g/mol

Ethyl hydrogen malonate is used as a reactant for the preparation of tetramic acids through Dieckmann ring closure and organocatalytic decarboxylative Doebner-Knoevenagel reactions. It is involved in the acylation reactions and Knoevenagel condensation with aldehydes. It is also used in the preparation of gamma-lactones from olefins by intermolecular carbolactonization in presence of Mn(III) acetate as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hydrogen malonate is used as a reactant for the preparation of tetramic acids through Dieckmann ring closure and organocatalytic decarboxylative Doebner-Knoevenagel reactions. It is involved in the acylation reactions and Knoevenagel condensation with aldehydes. It is also used in the preparation of gamma-lactones from olefins by intermolecular carbolactonization in presence of Mn(III) acetate as a catalyst.

Solubility
Miscible with water, chloroform and other solvents.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1071-46-1
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00020490
InChI Key HGINADPHJQTSKN-UHFFFAOYSA-N
Synonym ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate
PubChem CID 70615
ChEBI CHEBI:86907
IUPAC Name 3-ethoxy-3-oxopropanoic acid
SMILES CCOC(=O)CC(=O)O

Specifications

Density 1.119
Boiling Point 106°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4295
Linear Formula HO2CCH2CO2CH2CH3
Quantity 250 g
Beilstein 1758845
Solubility Information Miscible with water,chloroform and other solvents.
Formula Weight 132.12
Percent Purity 96%
Chemical Name or Material Ethyl hydrogen malonate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-992-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only