missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrazinoacetate hydrochloride, 97%

Catalog No. AC433150100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC433150100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC433150100 Supplier Thermo Scientific Chemicals Supplier No. 433150100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6945-92-2 | C4H11ClN2O2 | 154.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6945-92-2
Molecular Formula C4H11ClN2O2
Molecular Weight (g/mol) 154.59
MDL Number MFCD00012923
InChI Key HZZRIIPYFPIKHR-UHFFFAOYSA-N
Synonym ethyl hydrazinoacetate hydrochloride, ethyl 2-hydrazinylacetate hydrochloride, ethyl hydrazinoacetate hcl, hydrazino-acetic acid ethyl ester hcl, 2-hydrazinyl acetic acid ethyl ester hydrochloride, ethyl 2-hydrazinylacetate hcl, ethyl 2-hydrazinoacetate hydrochloride, acetic acid, hydrazino-, ethyl ester, monohydrochloride, acetic acid, ethyl ester, monohydrochloride, c4h10n2o2.clh
PubChem CID 11593468
IUPAC Name ethyl 2-hydrazinylacetate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)CNN

Specifications

Melting Point 147.0°C to 149.0°C
Color Colorless to Yellow
Elemental Analysis 29.82 to 32.31 % C 6.88 to 7.46 % H 17.39 to 18.84 % N
Infrared Spectrum Authentic
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NNHCH2CO2C2H5·HCl
Quantity 10 g
Formula Weight 154.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Ethyl hydrazinoacetate hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 230-104-3

RUO – Research Use Only