missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hydrazinoacetate hydrochloride, 97%

Catalog No. AAH6004214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6004214 25 g
AAH6004206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6004214 Supplier Thermo Scientific Chemicals Supplier No. H6004214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6945-92-2 | C4H11ClN2O2 | 154.59 g/mol

Ethyl hydrazinoacetate hydrochloride has been used in the preparation of three novel copper(II) complexes as condensation derivatives of 2-pyridinecarboxaldehyde, 2-acetylpyridine and 2-quinolinecarboxaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hydrazinoacetate hydrochloride has been used in the preparation of three novel copper(II) complexes as condensation derivatives of 2-pyridinecarboxaldehyde, 2-acetylpyridine and 2-quinolinecarboxaldehyde.

Notes
Store in cool dry conditions. Keep away from oxidizing agents.

Chemical Identifiers

CAS 6945-92-2
Molecular Formula C4H11ClN2O2
Molecular Weight (g/mol) 154.59
MDL Number MFCD00012923
InChI Key HZZRIIPYFPIKHR-UHFFFAOYSA-N
Synonym ethyl hydrazinoacetate hydrochloride, ethyl 2-hydrazinylacetate hydrochloride, ethyl hydrazinoacetate hcl, hydrazino-acetic acid ethyl ester hcl, 2-hydrazinyl acetic acid ethyl ester hydrochloride, ethyl 2-hydrazinylacetate hcl, ethyl 2-hydrazinoacetate hydrochloride, acetic acid, hydrazino-, ethyl ester, monohydrochloride, acetic acid, ethyl ester, monohydrochloride, c4h10n2o2.clh
PubChem CID 11593468
IUPAC Name ethyl 2-hydrazinylacetate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)CNN

Specifications

Melting Point 152°C to 154°C
Linear Formula H2NNHCH2CO2C2H5·HCl
Quantity 25 g
Beilstein 3912112
Formula Weight 154.6
Percent Purity 97%
Chemical Name or Material Ethyl hydrazinoacetate hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-104-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only