missing translation for 'onlineSavingsMsg'
Learn More

Ethyl hexafluoroglutaryl chloride, 97%, Thermo Scientific Chemicals

Catalog No. AAL1404906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1404906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1404906 Supplier Thermo Scientific Chemicals Supplier No. L1404906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18381-53-8 | Molecular Formula: C7H5ClF6O3 | Molecular Weight: 286.55 g/mol

Ethyl hexafluoroglutaryl chloride is used as a derivatising agent in gas chromatography. It is an active reagent used for the quantization of amphetamine and methamphetamine in urine by gas chromatography/mass spectroscopy (GC/MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl hexafluoroglutaryl chloride is used as a derivatising agent in gas chromatography. It is an active reagent used for the quantization of amphetamine and methamphetamine in urine by gas chromatography/mass spectroscopy (GC/MS).

Solubility
Miscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, bases and water.

Chemical Identifiers

CAS 18381-53-8
Molecular Formula C7H5ClF6O3
Molecular Weight (g/mol) 286.55
MDL Number MFCD00054671
InChI Key OLRXGDHRDQKNGW-UHFFFAOYSA-N
PubChem CID 161251
IUPAC Name ethyl 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxopentanoate
SMILES CCOC(=O)C(F)(F)C(F)(F)C(F)(F)C(Cl)=O

Specifications

Density 1.47
Boiling Point 158°C to 160°C
Refractive Index 1.359
Quantity 5 g
UN Number UN3265
Beilstein 1884844
Sensitivity Moisture sensitive
Synonym 4-carbethoxyhexafluorobutyryl chloride, ethyl hexafluoroglutaryl chloride, pentanoic acid, 5-chloro-2,2,3,3,4,4-hexafluoro-5-oxo-, ethyl ester, 4ce-hfba, 4-carbethoxyhexafluorobutyryl, ethyl hexafluoroglutaryl chloride 250mg, 4-ethoxycarbonyl-4,4,3,3,2,2-hexafluorobutyryl chloride, pentanoic acid,5-chloro-2,2,3,3,4,4-hexafluoro-5-oxo-, ethyl ester
Solubility Information Miscible with water.
Formula Weight 286.56
Percent Purity 97%
Chemical Name or Material Ethyl hexafluoroglutaryl chloride
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only