missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Heptanoate 98.0+%, TCI America™

Catalog No. p-7130297
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H003125ML 25 mL
H0031500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H003125ML Supplier TCI America Supplier No. H003125ML
Only null left

Chemical Identifiers

CAS 106-30-9
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00009538
InChI Key TVQGDYNRXLTQAP-UHFFFAOYSA-N
Synonym ethyl enanthate, oenanthic ether, wine oil, ethyl heptylate, grape oil, ethyl heptoate, heptanoic acid, ethyl ester, cognac oil, enanthylic ether, ethyl oenanthate
PubChem CID 7797
ChEBI CHEBI:86618
IUPAC Name ethyl heptanoate
SMILES CCCCCCC(=O)OCC

Specifications

Color Yellow
Boiling Point 188°C
Quantity 25 mL
Formula Weight 158.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Heptanoate
EINECSNumber (2)-0782
RTECSNumber MJ2087000
TSCA Yes