missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Heptanoate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 188°C |
| Quantity | 500 mL |
| Formula Weight | 158.24 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethyl Heptanoate |
Safety and Handling
| EINECSNumber | (2)-0782 |
| RTECSNumber | MJ2087000 |
| TSCA | Yes |