missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Heptadecanoate 97.0+%, TCI America™
$188.27 - $564.81
Chemical Identifiers
| CAS | 14010-23-2 |
|---|---|
| Molecular Formula | C19H38O2 |
| Molecular Weight (g/mol) | 298.511 |
| MDL Number | MFCD00026658 |
| InChI Key | KNXMUFRWYNVISA-UHFFFAOYSA-N |
| Synonym | Ethyl Margarate, Heptadecanoic Acid Ethyl Ester, Margaric Acid Ethyl Ester |
| PubChem CID | 26397 |
| IUPAC Name | ethyl heptadecanoate |
| SMILES | CCCCCCCCCCCCCCCCC(=O)OCC |
Chemical Identifiers
| 14010-23-2 | |
| 298.511 | |
| KNXMUFRWYNVISA-UHFFFAOYSA-N | |
| 26397 | |
| CCCCCCCCCCCCCCCCC(=O)OCC |
| C19H38O2 | |
| MFCD00026658 | |
| Ethyl Margarate, Heptadecanoic Acid Ethyl Ester, Margaric Acid Ethyl Ester | |
| ethyl heptadecanoate |
Specifications
| 14010-23-2 | |
| 200°C | |
| MFCD00026658 | |
| Ethyl Margarate, Heptadecanoic Acid Ethyl Ester, Margaric Acid Ethyl Ester | |
| CCCCCCCCCCCCCCCCC(=O)OCC | |
| 298.511 | |
| 298.51 | |
| Crystals |
| White | |
| C19H38O2 | |
| 5 g | |
| KNXMUFRWYNVISA-UHFFFAOYSA-N | |
| ethyl heptadecanoate | |
| 26397 | |
| ≥97.0% (GC) | |
| Ethyl Heptadecanoate |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : No