missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Glutaryl Chloride 98.0+%, TCI America™
$67.38 - $186.74
Chemical Identifiers
| CAS | 5205-39-0 |
|---|---|
| Molecular Formula | C7H11ClO3 |
| Molecular Weight (g/mol) | 178.612 |
| MDL Number | MFCD00013659 |
| InChI Key | KKJAQUGGQMCNJY-UHFFFAOYSA-N |
| Synonym | ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester |
| PubChem CID | 78881 |
| IUPAC Name | ethyl 5-chloro-5-oxopentanoate |
| SMILES | CCOC(=O)CCCC(=O)Cl |
Chemical Identifiers
| 5205-39-0 | |
| 178.612 | |
| KKJAQUGGQMCNJY-UHFFFAOYSA-N | |
| 78881 | |
| CCOC(=O)CCCC(=O)Cl |
| C7H11ClO3 | |
| MFCD00013659 | |
| ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester | |
| ethyl 5-chloro-5-oxopentanoate |
Specifications
| 5205-39-0 | |
| 108°C | |
| MFCD00013659 | |
| 3265 | |
| KKJAQUGGQMCNJY-UHFFFAOYSA-N | |
| ethyl 5-chloro-5-oxopentanoate | |
| 78881 | |
| ≥98.0% (T) | |
| Ethyl Glutaryl Chloride |
| Yellow | |
| C7H11ClO3 | |
| 5 g | |
| ethyl 4-chloroformyl butyrate, ethyl 5-chloro-5-oxovalerate, glutaric acid monoethyl ester chloride, pentanoic acid, 5-chloro-5-oxo-, ethyl ester, ethyl 4-chlorocarbonyl butanoate, pubchem16647, ksc939c3b, gamma-carbethoxybutyryl chloride, glutaric acid monoethylester chloride, 5-chloro-5-oxovaleric acid ethyl ester | |
| CCOC(=O)CCCC(=O)Cl | |
| 178.612 | |
| 178.61 | |
| Liquid |
Safety and Handling
TSCA : No