missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Gallate 98.0+%, TCI America™

Catalog No. g001625g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
G001625G 25 g
G0016100G 100 g
G0016500G 500 g
3 options

Catalog No. G001625G

Supplier: TCI America G001625G

Only null left

Chemical Identifiers

CAS 831-61-8
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.174
MDL Number MFCD00016430
InChI Key VFPFQHQNJCMNBZ-UHFFFAOYSA-N
Synonym ethyl gallate, gallic acid ethyl ester, phyllemblin, nipagallin a, progallin a, ethylgallate, nipa no. 48, gallic acid, ethyl ester, benzoic acid, 3,4,5-trihydroxy-, ethyl ester, nipa 48
PubChem CID 13250
ChEBI CHEBI:87247
IUPAC Name ethyl 3,4,5-trihydroxybenzoate
SMILES CCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 153°C
Quantity 25 g
Formula Weight 198.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Gallate
EINECSNumber (3)-1583
RTECSNumber LW7700000
TSCA Yes