missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Formate 98.0+%, TCI America™

Catalog No. F0053500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0053500ML 500 mL
F005325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0053500ML Supplier TCI America Supplier No. F0053500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-94-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00003294
InChI Key WBJINCZRORDGAQ-UHFFFAOYSA-N
Synonym areginal, formic acid, ethyl ester, ethyl methanoate, formic acid ethyl ester, aethylformiat, ethyl formic ester, ethylformate, ethylformiaat, formic ether, ethylformiat
PubChem CID 8025
ChEBI CHEBI:52342
IUPAC Name ethyl formate
SMILES CCOC=O

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 54°C
Quantity 500 mL
UN Number 1190
Formula Weight 74.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Formate
EINECSNumber (2)-0678
RTECSNumber LQ8400000
TSCA Yes