missing translation for 'onlineSavingsMsg'
Learn More

Ethyl fluoroacetate, 98%

Catalog No. AAH6134009
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6134009 10 g
AAH6134018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6134009 Supplier Thermo Scientific Chemicals Supplier No. H6134009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 459-72-3 | C4H7FO2 | 106.096 g/mol

Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl fluoroacetate is used in the synthesis of 4-deoxy-4-fluoro-muscarines. It is also used as starting reagent for the synthesis of ethyl(diethoxyphosphoryl)fluoroacetate. and in development of an ammonolysis-based microencapsulation process.

Solubility
Decomposes in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 459-72-3
Molecular Formula C4H7FO2
Molecular Weight (g/mol) 106.096
MDL Number MFCD00000450
InChI Key VCYZVXRKYPKDQB-UHFFFAOYSA-N
Synonym ethyl fluoroacetate, ethyl monofluoroacetate, acetic acid, fluoro-, ethyl ester, fluoroacetic acid ethyl ester, unii-aus1l6um7b, ethylester kyseliny fluoroctove, monofluoroacetic acid ethyl ester, aus1l6um7b, ethylester kyseliny fluoroctove czech, acetic acid, 2-fluoro-, ethyl ester
PubChem CID 9988
IUPAC Name ethyl 2-fluoroacetate
SMILES CCOC(=O)CF

Specifications

Color Colorless to Yellow
Density 1.093 g/mL
Boiling Point 119°C to 120°C
Refractive Index 1.375
Linear Formula FCH2CO2CH2CH3
Quantity 10 g
UN Number UN2929
Beilstein 1743761
Merck Index 14,4167
Solubility Information Decomposes in water.
Formula Weight 106.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl fluoroacetate
Show More Show Less
Hazard Category H226-H300+H310+H330
Hazard Statement GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 207-297-8
RTECSNumber AH7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only