missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (ethoxymethylene)cyanoacetate, 98%

Catalog No. AAA1236618
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1236618 50 g
AAA1236630 250 g
AAA123660B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1236618 Supplier Thermo Scientific Chemicals Supplier No. A1236618
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 94-05-3 | C8H11NO3 | 169.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94-05-3
Molecular Formula C8H11NO3
Molecular Weight (g/mol) 169.18
MDL Number MFCD00009136
InChI Key KTMGNAIGXYODKQ-SREVYHEPSA-N
Synonym ethyl ethoxymethylene cyanoacetate, e-ethyl 2-cyano-3-ethoxyacrylate, ethyl 2-cyano-3-ethoxyacrylate, ethyl 3-ethoxy-2-cyanoacrylate, ethyl 2e-2-cyano-3-ethoxyprop-2-enoate, ethyl cyano ethoxymethylene acetate, 2-propenoic acid, 2-cyano-3-ethoxy-, ethyl ester, 2-cyano-3-ethoxy-2-propenoic acid ethyl ester, 2-cyano-3-ethoxyacrylic acid ethyl ester, ethyl 2e-2-cyano-3-ethoxy-2-propenoate
PubChem CID 1715183
IUPAC Name ethyl (E)-2-cyano-3-ethoxyprop-2-enoate
SMILES CCO\C=C(\C#N)C(=O)OCC

Specifications

Melting Point 47°C to 52°C
Boiling Point 190°C to 191°C (30 mmHg)
Flash Point 130°C (266°F)
Quantity 50 g
UN Number UN3439
Beilstein 878814
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material Ethyl (ethoxymethylene)cyanoacetate
Hazard Category H302-H315-H317-H318-H334-H335
Hazard Statement GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 202-299-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only