missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Ether (Spectranalyzed™), Fisher Chemical™

Catalog No. p-215987
Change view
Click to view available options
Quantity:
4 L
Packaging:
Tin Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
E1974 4 L Tin Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E1974 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. FLE1974
Only null left

Chemical Identifiers

CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC

Specifications

Melting Point -116°C
Absorbance 0.01 AU max. at 400nm, 0.03 AU max. at 270nm, 0.1 AU max. at 254nm, 0.2 AU max. at 242nm, 0.4 AU max. at 231nm, 0.6 AU max. at 224nm, 0.8 AU max. at 221nm, 1 AU max. at 220nm
Color Colorless
Acidity 5ppm max.
Density 0.714g/cm³
Quantity 4 L
Flash Point -45°C (-113°F)
Identification Pass Test
Packaging Tin Can
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range ≥99 %
Vapor Pressure 587mbar at 20°C
Linear Formula C2H5OC2H5
Viscosity 0.2448 cP at 20°C
UN Number 1155
Boiling Point 34.6°C
Formula Weight 74.12
Percent Purity ≥99%
Grade Spectranalyzed
Chemical Name or Material Ethyl Ether
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Extremely flammable liquid and vapor. Vapor may cause flash fire. Harmful if swallowed. Irritating to eyes and skin. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. May cause irritation of respiratory tract. Repeated exposure may cause skin dryness or cracking. Hygroscopic. Wear personal protective equipment. Handle under an inert atmosphere. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid contact with skin, eyes and clothing. Do not breathe vapors or spray mist.

NFPA

Health:1
Flammability:4
Instability:1
DOTInformation DOT Class 3, : Flammable Liquid